Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium citrate monohydrate 99.0-100.5% (by perchloric acid titration), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E332

Supplier: MilliporeSigma
Synonyms: Tripotassium citrate monohydratePotassium citrate tribasic monohydrateCitric acid tripotassium salt monohydrateTripotassium 2-hydroxypropane-1,2,3-tricarboxylate monohydrate

1.04956.5000 1.04956.9029 1.04956.9029
EM1.04956.9029EA 2480 USD InStock
EM1.04956.9029 CA1.04956.5000 CA1.04956.9029
Potassium citrate monohydrate 99.0-100.5% (by perchloric acid titration), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, FCC, E332
Potassium citrate monohydrate
Elemental impurity specifications have been set considering ICH Q3D (guideline for elemental impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Corresponds to Ph Eur, BP, USP, FCC, E 332

Certifications: Conforms to the purity criteria on food additives according to the current European Commission Regulation.


Formula: HOC(COOK)(CH₂COOK)₂·H₂O
MW: 324.41 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150442
CAS Number: 6100-05-6
EINECS: 212-755-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/37/2297237.jpg
Order Now
Specification Test Results
Assay (Perchloric Acid Titration, Calculated on Dried Substance) 99.0 - 100.5%
Assay (Perchloric Acid Titration After Drying) 99.0 - 100.5%
Identity Passes Test
Appearance of Solution Passes Test
Alkalinity Passes Test
Acidity or Alkalinity Passes Test
pH-Value (5%; Water) 7.5 - 9.0
Chloride (Cl) ≤0.0050%
Sulfate (SO₄) ≤0.0150%
Heavy Metals (As Pb) ≤0.0005%
Al (Aluminium) ≤0.0001%
As (Arsenic) ≤0.0001%
Hg (Mercury) ≤0.0001%
Na (Sodium) ≤0.3%
Pb (Lead) ≤0.0001%
Oxalate (C₂O₄) ≤0.009%
Tartrate (C₄H₄O₆) Passes Test
Residual Solvents (CH Q3C) Excluded by Manufacturing Process
Readily Carbonisable Substance Passes Test
Loss On Drying (180 °C) 4.0 - 6.0%
Water 4.0 - 7.0%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty