Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide, pellets, pure, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Potassium hydrateCaustic potash

1.05012.5000 1.05012.1000 1.05012.1000 1.05012.9050 1.05012.5000 1.05012.9050
EM1.05012.9050EA 1431 USD InStock
EM1.05012.9050 EM1.05012.1000 EM1.05012.5000 CA1.05012.9050 CA1.05012.1000 CA1.05012.5000
Potassium hydroxide, pellets, pure, Supelco®
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320 °C (1013 hPa)
Melting Pt: 360 °C
Density: 2.04 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 1813
ADR: 8,II
REACH: 01-2119487136-33
Merck Index: 12,07806
Order Now
Specification Test Results
Solubility 1130 g/l (20 °C)
Melting Point 360 °C
Molar Mass 56.11 g/mol
Boiling Point 1320 °C
Vapor Pressure (20 °C)
Density 2.04 g/cm3 (20 °C)
pH 14 (56 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty