Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium hydrogen phosphate, anhydrous, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Synonyms: di-Potassium hydrogen orthophosphateDibasic Potassium PhosphatePotassium phosphate dibasicPotassium Phosphate, Dibasic, (dipotassium hydrogen phosphate)Potassium hydrogen phosphateDipotassium hydrogen phosphate

1.05109.0500 1.05109.0500 1.05109.0100 1.05109.0100
EM1.05109.0500CS 20391 USD InStock
EM1.05109.0500 CA1.05109.0100 CA1.05109.0500 EM1.05109.0100
di-Potassium hydrogen phosphate, anhydrous, Suprapur® for trace metal analysis, Supelco®
di-Potassium hydrogen phosphate


Formula: K₂HPO₄
MW: 174.18 g/mol
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: 253 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-913-4
REACH: 01-2119490224-41
Merck Index: 12,07829
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/42/16640642.jpg
Order Now
Specification Test Results
Solubility 1600 g/l (20 °C)
Molar Mass 174.18 g/mol
Bulk Density 700 - 1000 kg/m3
Density 2.44 g/cm3 (20 °C)
pH 9 (10 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty