Would you like to log in again?
SABOURAUD 4% Dextrose Agar, MilliporeSigma
Supplier: MilliporeSigma
1.05438.5000 1.05438.5000
EM1.05438.5000EA 1429.49 USD InStock
CA1.05438.5000 EM1.05438.5000
SABOURAUD 4% Dextrose Agar, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
  • Clear, yellowish-brown appearance
  • Fair to very good growth in Trichophyton mentagrophytes ATCC 18748
  • Fair to good growth in Trichophyton rubrum ATCC 28188
  • Fair to good growth in Trichophyton ajelloi ATCC 28454
  • Fair to very good growth in Microsporum gallinae ATCC 12108
  • Good to very good growth in Microsporum canis ATCC 36299
  • Good to very good growth in Geotrichum candidum DSM 1240
  • Good to very good growth in Penicillium commune ATCC 10428
In North America 1.05438.5000 replaces and is identical to 1.05438.5007
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty