Would you like to log in again?
Tryptic Soy Broth, MilliporeSigma
Supplier: MilliporeSigma
1.05459.9025 1.05459.9025
EM1.05459.9025EA 3290.56 USD InStock
CA1.05459.9025 EM1.05459.9025
Tryptic Soy Broth, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Casein-peptone soymeal-peptone broth for microbiology (According harm. EP/USP/JP and ISO). Pepton from casein 17.0; Pepton from soymeal 3.0; D(+)-Glucose monohydrate 2.5; Sodium chloride 5.0; di-Potassium hydrogen phosphate 2.5.
  • Clear after seven-day stability test at room temperature
  • 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • 10 to 100 inoculum on reference medium Bacillus subtilis ATCC 6633 (WDCM 00003)
  • 10 to 100 inoculum on reference medium Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)
  • 10 to 100 inoculum on reference medium Escherichia coli ATCC 8739 (WDCM 00012)
  • 10 to 100 inoculum on reference medium Streptococcus pneumoniae ATCC 6301
  • 10 to 100 inoculum on reference medium Salmonella typhimurium ATCC 14028 (WDCM 00031)
  • 10 to 100 inoculum on reference medium Staphylococcus aureus ATCC 25923 (WDCM 00034)
  • 10 to 100 inoculum on reference medium Staphylococcus epidermidis ATCC 12228 (WDCM 00036)
  • 10 to 100 inoculum on reference medium Candida albicans ATCC 2091 (WDCM 00055)
  • 10 to 100 inoculum on reference medium Candida albicans ATCC 10231 (WDCM 00054)
  • 10 to 100 inoculum on reference medium Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)
  • Visible growth in 18 to 24 hours at 30 to 35°C in Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • Visible growth in 18 to 24 hours at 30 to 35°C in Bacillus subtilis ATCC 6633 (WDCM 00003)
  • Visible growth in 18 to 24 hours at 30 to 35°C in Pseudomonas aeruginosa ATCC 9027 (WDCM 00026)
  • Visible growth in 18 to 24 hours at 30 to 35°C in Escherichia coli ATCC 8739 (WDCM 00012)
  • Visible growth in 18 to 24 hours at 30 to 35°C in Streptococcus pneumoniae ATCC 6301
  • Visible growth in 18 to 24 hours at 30 to 35°C in Salmonella typhimurium ATCC 14028 (WDCM 00031)
  • Visible growth in 3 days at 20 to 25°C in Staphylococcus aureus ATCC 6538 (WDCM 00032)
  • Visible growth in 3 days at 20 to 25°C in Staphylococcus aureus ATCC 25923 (WDCM 00034)
  • Visible growth in 3 days at 20 to 25°C in Staphylococcus epidermidis ATCC 12228 (WDCM 00036)
  • Visible growth in 3 days at 20 to 25°C in Bacillus subtilis ATCC 6633 (WDCM 00003)
  • Visible growth in 5 days at 20 to 25°C in Candida albicans ATCC 2091 (WDCM 00055)
  • Visible growth in 5 days at 20 to 25°C in Candida albicans ATCC 10231 (WDCM 00054)
  • Visible growth in 5 days at 20 to 25°C in Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty