Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 21% in aqueous solution analytical reagent, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

1.05593.9025 1.05593.9025
EM1.05593.9025EA 900 USD InStock
EM1.05593.9025 CA1.05593.9025
Sodium hydroxide 21% in aqueous solution analytical reagent, Supelco®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Density 1.219 g/cm3 (20 °C)
pH 11 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty