Would you like to log in again?
Aluminium Oxide F254 Plates And Sheets, MilliporeSigma
Supplier: MilliporeSigma
1.05731.0001 1.16717.0001 1.16717.0001 1.05713.0001 1.05581.0001 1.05550.0001
EM1.05550.0001EA 584.75 USD InStock
CA1.16717.0001 EM1.05550.0001 EM1.05713.0001 EM1.05731.0001 10143-276 EM1.05581.0001
Aluminium Oxide F254 Plates And Sheets, MilliporeSigma
TLC Plates
Aluminium oxide plates are used to analyze basic and neutral compounds at different pH levels
MilliporeSigma # 1.05550.0001 replaces and is identical to MilliporeSigma # 5550-7.
MilliporeSigma # 1.05551.0001 replaces and is identical to MilliporeSigma # 5551-7.
MilliporeSigma # 1.05581.0001 replaces and is identical to MilliporeSigma # 5581-7.
MilliporeSigma # 1.05713.0001 replaces and is identical to MilliporeSigma # 5713-7.
MilliporeSigma # 1.05731.0001 replaces and is identical to MilliporeSigma # 5731-3.
Depending on the pH range in question, professionals can choose between two kinds of aluminum oxide plates. Under aqueous conditions, basic aluminum oxide plates are best for separating basic compounds, while neutral plates are ideal for separating neutral compounds. Available with or without a fluorescence indicator, these TLC aluminum plates handle a wide variety of applications thanks to neutral and basic aluminum oxides with 60Å and 150Å pore sizes.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty