Would you like to log in again?
OGYE Agar Base, MilliporeSigma
Supplier: MilliporeSigma
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
1.05978.0500 1.05978.0500
EM1.05978.0500EA 284.52 USD InStock
EM1.05978.0500 CA1.05978.0500
OGYE Agar Base, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
Oxytetracycline-glucose-yeast extract agar is used for the selective isolation and enumeration of yeasts and moulds in foods.
  • pH: 6.6 ±0.2 at 25 °C
The base medium allows good growth of yeasts and moulds. Oxytetracycline inhibits the growth of bacteria.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty