Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methanol, SupraSolv® for GC-ECD/FID, Supelco®

Supplier: MilliporeSigma

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: Hydroxy methaneMethyl hydroxideMethyl alcoholMethylolCarbinolWood alcohol

1.06011.2500 1.06011.2500 1.06011.1000 1.06011.1000 1.06011.4000 1.06011.4000
EM1.06011.4000EA 237 USD InStock
EM1.06011.4000 CA1.06011.2500 EM1.06011.2500 CA1.06011.1000 EM1.06011.1000 CA1.06011.4000
Methanol, SupraSolv® for GC-ECD/FID, Supelco®
Methanol
SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.


Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3,II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://us-legacy.vwr.com/stibo/web/std.lang.all/33/12/733312.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -98 °C
Molar Mass 32.04 g/mol
Boiling Point 64.5 °C (1013 hPa)
Vapor Pressure 128 hPa (20 °C)
Flash Point 10 °C
Refractive Index 1.33
Explosion Limit 5.5 - 44 %(V)
Density 0.792 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 455 °C DIN 51794
Evaporation number 6.3
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty