Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Methane dichlorideMethylene chloride

1.06050.6025 1.06050.2500 1.06050.4000 1.06050.4000 1.06050.6010 1.06050.9190 1.06050.9190 1.06050.6025 1.06050.6010 1.06050.1000 1.06050.2500
EM1.06050.6025EA 1819 USD InStock
EM1.06050.6025 EM1.06050.4000 EM1.06050.9190 CA1.06050.6010 CA1.06050.9190 CA1.06050.4000 EM1.06050.2500 CA1.06050.6025 CA1.06050.1000 EM1.06050.6010 CA1.06050.2500
Dichloromethane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Dichloromethane


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Solubility 20 g/l (20 °C)
Melting Point -95 °C
Molar Mass 84.93 g/mol
Boiling Point 40 °C (1013 hPa)
Vapor Pressure 475 hPa (20 °C)
Refractive Index 1.42
Explosion Limit 13 - 22 %(V)
Density 1.33 g/cm3 (20 °C)
pH (H2O, 20 °C) neutral
Ignition Point 605 °C DIN 51794
Evaporation number 1.9

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty