Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Dichloromethane ≥99.8% stabilized, SupraSolv® for GC-ECD/FID, distilled, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Methane dichlorideMethylene chloride

Stabilization: Stabilised with 2-methyl-2-butene (iso-amylene)

1.06054.1000 1.06054.4000 1.06054.4000 1.06054.1000 1.06054.2500 1.06054.2500
EM1.06054.2500CS 1184 USD InStock
CA1.06054.4000 EM1.06054.2500 EM1.06054.1000 CA1.06054.1000 EM1.06054.4000 CA1.06054.2500
Dichloromethane ≥99.8% stabilized, SupraSolv® for GC-ECD/FID, distilled, Supelco®
Dichloromethane
SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. Typical applications include the determination of polychlorinated biphenyls (PCB) in water and soil or pesticides in fruits and vegetables. Also ideal for many EPA protocols.


Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://us-legacy.vwr.com/stibo/web/std.lang.all/14/30/20961430.jpg
Order Now
Specification Test Results
Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue = 5.0 mg/l
Water = 0.01%
Color =10 Hazen
GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) = 3pg/ml
GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty