Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Naphthol, GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: α-Naphtholα-Hydroxynaphthalene1-Hydroxynaphthalene

1.06223.0050 1.06223.0250 1.06223.0250 1.06223.0050
EM1.06223.0050CS 1874 USD InStock
CA1.06223.0050 EM1.06223.0050 EM1.06223.0250 CA1.06223.0250
1-Naphthol, GR for analysis, Supelco®
1-Naphthol


Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 94 °C
Density: 1.224 g/cm³ (20 °C)
Flash Pt: 125 °C
Storage Temperature: Ambient
MDL Number: MFCD00003930
CAS Number: 90-15-3
EINECS: 201-969-4
UN: 2811
ADR: 6.1,III
Merck Index: 12,06470
Order Now
Specification Test Results
Solubility 0.1 g/l (20 °C)
Melting Point 94 - 96 °C
Molar Mass 144.17 g/mol
Bulk Density 450 kg/m3
Boiling Point 288 °C (1013 hPa)
Vapor Pressure 1.3 hPa (94 °C)
Flash Point 125 °C
Density 1.28 g/cm3 (20 °C)
Ignition Point 510 °C
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty