Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥97% (NaOH basis by acidimetry), pellets, pure, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

1.06462.5000 1.06462.1000 1.06462.9050 1.06462.5000 1.06462.9050
EM1.06462.5000CS 1590 USD InStock
CA1.06462.9050 CA1.06462.1000 CA1.06462.5000 EM1.06462.5000 EM1.06462.9050
Sodium hydroxide ≥97% (NaOH basis by acidimetry), pellets, pure, Supelco®
Sodium hydroxide
1.06462.1000 is offered as an alternative to SX0600-1. Different Pack Size, Same Specifications.
1.06462.9050 is offered as an alternative to SX0600-30. Different Pack Size, Same Specifications.


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 323 °C
Density: 2.13 g/mL (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Solubility 1090 g/l (20 °C)
Melting Point 323 °C
Molar Mass 40.00 g/mol
Boiling Point 1390 °C (1013 hPa)
Vapor Pressure (20 °C)
Density 2.13 g/cm3 (20 °C)
pH 14 (50 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty