Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide ≥99% (NaOH basis by acidimetry), pellets, EMSURE® ISO for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

M-Clarity™ quality level = MQ300

1.06498.9050 1.06498.9025 1.06498.1000 1.06498.5000 1.06498.0500 1.06498.0500 1.06498.1000 1.06498.5000 1.06498.9050 1.06498.9025
EM1.06498.9025EA 1125 USD InStock
EM1.06498.9025 CA1.06498.9025 EM1.06498.0500 CA1.06498.1000 EM1.06498.9050 EM1.06498.1000 CA1.06498.5000 CA1.06498.0500 CA1.06498.9050 EM1.06498.5000
Sodium hydroxide ≥99% (NaOH basis by acidimetry), pellets, EMSURE® ISO for analysis, Supelco®
Sodium hydroxide


Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C (1013 hPa)
Melting Pt: 318 °C (lit.)
Density: 2.13 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II
REACH: 01-2119457892-27
Merck Index: 14,08627
Order Now
Specification Test Results
Assay (acidimetric, NaOH) 99% min
Carbonate (as Na₂CO₃) 1.0% max
Chloride (Cl) 0.012% max
Phosphate (PO₄) 0.0005% max
Silicate (SiO₂) 0.001% max
Sulfate (SO₄) 0.010% max
Total nitrogen (N) 0.0003% max
Heavy metals (as Pb) 0.0005% max
Al (Aluminium) 0.0005% max
As (Aresnic) 0.0001% max
Ca (Calcium) 0.0005% max
Cu (Copper) 0.0002% max
Fe (Iron) 0.0005% max
K (Pottassium) 0.05% max
Mg (Magnesium) 0.0005% max
Ni (Nickel) 0.00025% max
Pb (Lead) 0.0005% max
Zn (Zinc) 0.001% max
. .
. .
. .

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty