Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium thiosulfate, anhydrous, Supelco®

Supplier: MilliporeSigma
Synonyms: di-Sodium thiosulfateSodium hyposulphitedi-Sodium thiosulphateSodiumthiosulfateSodium hyposulfite

M-Clarity™ quality level = MQ200

1.06512.2500 1.06512.9025 1.06512.9050 1.06512.0250 1.06512.0250 1.06512.9050 1.06512.2500 1.06512.9025
EM1.06512.9050EA 1495 USD InStock
CA1.06512.9025 CA1.06512.2500 EM1.06512.9050 CA1.06512.0250 EM1.06512.2500 CA1.06512.9050 EM1.06512.0250 EM1.06512.9025
Sodium thiosulfate, anhydrous, Supelco®
Sodium thiosulphate


Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 48 °C
Density: 1.66 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-867-5
Merck Index: 12,08844
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty