Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrasodium diphosphate decahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium pyrophosphate decahydratetetra-Sodium pyrophosphate decahydrateSodium pyrophosphate tetrabasic decahydrate

1.06591.2500 1.06591.0500 1.06591.9050 1.06591.0500 1.06591.2500 1.06591.9050
EM1.06591.9050EA 15543 USD InStock
EM1.06591.9050 EM1.06591.2500 CA1.06591.2500 EM1.06591.0500 CA1.06591.9050 CA1.06591.0500
Tetrasodium diphosphate decahydrate, EMSURE® for analysis, Supelco®
tetra-Sodium diphosphate decahydrate


Formula: Na₄P₂O₇·10H₂O
MW: 446.06 g/mol
Melting Pt: 990 °C
Density: 1.82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149200
CAS Number: 13472-36-1
EINECS: 231-767-1
Order Now
Specification Test Results
Solubility 20 g/l (20 °C) (anhydrous substance)
Melting Point 990 °C (anhydrous substance)
Molar Mass 446.06 g/mol
Bulk Density 650 kg/m3
Density 1.82 g/cm3 (20 °C)
pH 9.5 - 10.5 (50 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty