Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) chloride hexahydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Nickel dichloride hexahydrate

1.06717.1000 1.06717.0250 1.06717.0250 1.06717.1000
EM1.06717.0250CS 3758 USD InStock
EM1.06717.0250 EM1.06717.1000 CA1.06717.0250 CA1.06717.1000
Nickel(II) chloride hexahydrate, EMSURE® ACS for analysis, Supelco®
Nickel (II) chloride hexahydrate


Formula: NiCl₂·6H₂O
MW: 237.69 g/mol
Melting Pt: 140 °C
Density: 1.84 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 12,06588
Order Now
Specification Test Results
Solubility 553 g/l (20 °C)
Melting Point 1001 °C (anhydrous substance),decomposes
Molar Mass 237.66 g/mol
Bulk Density 640 kg/m3
Boiling Point 987 °C (sublimed),(anhydrous substance)
pH 4.9 (100 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic materials

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty