Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Papanicolaou's solution 2b Orange II solution stain for cytology, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ100

1.06887.2500
EM1.06887.2500EA 302.44 USD InStock
EM1.06887.2500
Papanicolaou's solution 2b Orange II solution stain for cytology, Sigma-Aldrich®
Papanicolaou's solution 2b Orange II solution
Cytoplasm staining of mature and keratinized cells in cytology

The cytological staining technique developed by Papanicolaou is still the standard method in cancer and hormone cycle diagnosis. The effect of the orange staining solution is considerable in smears with keratinized cells under acidic pH conditions, when the obvious orangeophilic cytoplasmic stain is recognizable and may point to the presence of hyperkeratosis, HPV infection or carcinoma cells. Papanicolaou's solution 2b, Orange II solution gives a intense reddish staining result with mature and keratinized squamous cells. Papanicolaou's solution 2a, Orange G solution gives a gentle yellow-orange staining result with mature and keratinized squamous cells. A 500 ml bottle is sufficient for 1500 to 2000 stainings. It is registered as IVD and CE product.


Density: 0.83 g/cm3 (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
UN: 1987
ADR: 3,II
IVDLogo
Order Now
Specification Test Results
Suitability for microscopy (Vaginal smear) passes test
Nuclei blue to dark violet
Cyanophilic cytoplasm blue-green
Eosinophilic cytoplasm pink

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty