Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

penta-Sodium triphosphate for BOD determination acc. to DIN 38409-H-51, Supelco®

Supplier: MilliporeSigma
Synonyms: Sodium triphosphate pentabasicSodium tripolyphosphate

1.06999.1000 1.06999.9025 1.06999.9025 1.06999.1000
EM1.06999.9025EA 740 USD InStock
EM1.06999.9025 CA1.06999.1000 EM1.06999.1000 CA1.06999.9025
penta-Sodium triphosphate for BOD determination acc. to DIN 38409-H-51, Supelco®
Pentasodium triphosphate


Formula: Na₅O₁₀P₃
MW: 367.86 g/mol
Melting Pt: 622 °C (dec.)
Density: 2.52 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003514
CAS Number: 7758-29-4
EINECS: 231-838-7
Merck Index: 12,08846
Order Now
Specification Test Results
Assay (acidimetric, P₂O₅) 57 - 59 %
pH-value (1 %; water) 43382
Fe (Iron) ≤ 0.005 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty