Would you like to log in again?
Oxford Listeria Selective Supplement, MilliporeSigma
Supplier: MilliporeSigma
1.07006.0010 1.07006.0010
EM1.07006.0010PK 200.72 USD InStock
CA1.07006.0010 EM1.07006.0010
Oxford Listeria Selective Supplement, MilliporeSigma
Media for Microbiology Additives and Supplements for Microbiology
Supplement tested in Oxford Listeria selective agar (base) art
  • Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 19111 (WDCM 00020)
  • Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 13932 (WDCM 00021)
  • Greater than or equal to 70% recovery on test medium Listeria monocytogenes ATCC 35152 (WDCM 00109)
  • Greater than or equal to 70% recovery on test medium Listeria innocua ATCC 33090 (WDCM 00017)
  • Less than or equal to 0.01% recovery on test medium Enterococcus faecalis ATCC 19433 (WDCM 00009)
  • Less than or equal to 0.01% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Black zone Listeria monocytogenes ATCC 19111 (WDCM 00020) +
  • Black zone Listeria monocytogenes ATCC 13932 (WDCM 00021) +
  • Black zone Listeria monocytogenes ATCC 35152 (WDCM 00109) +
  • Black zone Listeria innocua ATCC 33090 (WDCM 00017) +
  • Growth inhibited in Staphylococcus aureus ATCC 25923 (WDCM 00034)
  • Growth inhibited in Candida albicans ATCC 10231 (WDCM 00054)
  • Reference Tryptic soy agar
In North America 1.07006.0010 replaces and is identical to 1.07006.0017
no. 1.07004. Cycloheximide ( >= 90 % - <= 100 % ) 66-81-9 and acriflavine hydrochloride ( >= 1 % - < 5 % ) 69235-50-3.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty