Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Aminoantipyrine, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: 4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one4-Aminophenazone

1.07293.0010 1.07293.0100 1.07293.0010 1.07293.0100
EM1.07293.0100CS 10157 USD InStock
CA1.07293.0100 CA1.07293.0010 EM1.07293.0100 EM1.07293.0010
4-Aminoantipyrine, GR for analysis, Supelco®
4-Aminoantipyrine


Formula: C₁₁H₁₃N₃O
MW: 203.24 g/mol
Melting Pt: 107…109 °C
Storage Temperature: Ambient
MDL Number: MFCD00003145
CAS Number: 83-07-8
EINECS: 201-452-3
Merck Index: 13,00596
Order Now
Specification Test Results
Solubility 56 g/l (20 °C)
Melting Point 107 - 109 °C
Molar Mass 203.24 g/mol
Bulk Density 430 kg/m3
pH 7.1 (100 g/l, H2O, 20 °C) (slurry)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty