Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrogen peroxide 30%, Suprapur® for trace metal analysis, Supelco®

Supplier: Avantor
Danger

1.07298.1000 1.07298.0250 1.07298.0250 1.07298.1000
CA1.07298.1000CS 8914.66 USD InStock
CA1.07298.1000 CA1.07298.0250 EM1.07298.1000 EM1.07298.0250
Hydrogen peroxide 30%, Suprapur® for trace metal analysis, Supelco®
Hydrogen peroxide
EMD # 1.07298.0250 replaces and is identical to EMD # 7298-2.
EMD # 1.07298.1000 replaces and is identical to EMD # 7298-4.


Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 (8),II
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -26 °C
Boiling Point 107 °C (1013 hPa)
Vapor Pressure 18 hPa (20 °C)
Density 1.11 g/cm3 (20 °C)
pH 2 - 4 (H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty