Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexachloroplatinic(IV) acid ∼10% (3.8% Pt), GR analytical reagent, Supelco®

Supplier: MilliporeSigma
Danger

1.07341.0005 1.07341.0025 1.07341.0005 1.07341.0025
EM1.07341.0005CS 106026 USD InStock
EM1.07341.0005 EM1.07341.0025 CA1.07341.0005 CA1.07341.0025
Hexachloroplatinic(IV) acid ∼10% (3.8% Pt), GR analytical reagent, Supelco®
Hexachloroplatinic (IV) acid


Formula: H₂Cl₆Pt
MW: 409.81 g/mol
MDL Number: MFCD00011330
CAS Number: 16941-12-1
UN: 3264
ADR: 8,III
Order Now
Specification Test Results
Solubility (20 °C) soluble
Density 1.06 g/cm3 (20 °C)
pH 0.6 (H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty