Would you like to log in again?
Lactose TTC Agar with Tergitol® 7, MilliporeSigma
Supplier: MilliporeSigma
1.07680.0500
EM1.07680.0500EA 317.59 USD InStock
EM1.07680.0500
Lactose TTC Agar with Tergitol® 7, MilliporeSigma
Media for Microbiology Dehydrated Media for Microbiology
  • Clear, green appearance
  • Greater than or equal to 70% recovery on test medium Escherichia coli ATCC 25922 (WDCM 00013)
  • Greater than or equal to 70% recovery on test medium Citrobacter freundii ATCC 8090
  • Greater than or equal to 70% recovery on test medium Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)
  • Yellow medium color for Escherichia coli ATCC 25922 (WDCM 00013)
  • Yellow medium color for Citrobacter freundii ATCC 8090
  • Blue medium color for Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)
  • Yellow-orange colony color for Escherichia coli ATCC 25922 (WDCM 00013)
  • Yellow-orange colony color for Citrobacter freundii ATCC 8090
  • Red colony color for Pseudomonas aeruginosa ATCC 27853 (WDCM 00025)
  • Growth inhibited for Bacillus cereus ATCC 11778 (WDCM 00001)
Suitable for microbiology.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty