Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sand (sea sand), Extra Pure, Supelco®

Supplier: MilliporeSigma

M-Clarity™ quality level = MQ200

1.07711.1000 1.07711.9025 1.07711.9025 1.07711.5000
EM1.07711.9025EA 1842 USD InStock
CA1.07711.5000 CA1.07711.9025 CA1.07711.1000 EM1.07711.9025
Sand (sea sand), Extra Pure, Supelco®
Sea sand


Formula: SiO₂
MW: 60.08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2.65 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 14808-60-7
EINECS: 231-545-4
Order Now
Specification Test Results
Substances soluble in hydrochloric acid ≤ 0.5 %
Chloride (Cl) ≤ 0.01 %
Loss on ignition (900 °C) ≤ 0.5 %
Particle size (0.1 - 0.315 mm) ≥ 90 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty