Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin, granulated, EMSURE® ∼ 4 mm for analysis, Supelco®

Supplier: MilliporeSigma

1.07806.0250 1.07806.1000 1.07806.1000 1.07806.0250
EM1.07806.1000CS 28146 USD InStock
CA1.07806.1000 EM1.07806.1000 EM1.07806.0250 CA1.07806.0250
Tin, granulated, EMSURE® ∼ 4 mm for analysis, Supelco®
Tin


Formula: Sn
MW: 118.71 g/mol
MDL Number: MFCD00133862
CAS Number: 7440-31-5
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 232 °C
Molar Mass 118.71 g/mol
Boiling Point 2260 °C (1013 hPa)
Density 7.3 g/cm3 (20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty