Would you like to log in again?
Hydrazine Test, MColortest™, MilliporeSigma
Supplier: MilliporeSigma
MColortest™
1.08017.0001 1.08017.0001
EM1.08017.0001EA 150 USD InStock
CA1.08017.0001 EM1.08017.0001
Hydrazine Test, MColortest™, MilliporeSigma
Environmental Test Kits
The test can be used for the colorimetric determination of hydrazine in boiler water and boiler feed water with colour-matching vessel.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty