Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Potassium sodium tartrate tetrahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Synonyms: (R,R)-(+)-Tartaric acid potassium sodium salt tetrahydrateL(+)-Tartaric acid potassium sodium salt tetrahydratePotassium sodium tartrate tetrahydratePotassium sodium L(+)-tartrate tetrahydrate(+)-Potassium sodium L-tartrate tetrahydratePotassium sodium (2R,3R)-2,3-dihydroxysuccinate tetrahydrate(R,R)-(+)-Potassium sodium tartrate tetrahydrate

1.08087.1000 1.08087.1000 1.08087.0500 1.08087.9050 1.08087.5000 1.08087.0500 1.08087.5000 1.08087.9050
EM1.08087.5000CS 7646 USD InStock
EM1.08087.5000 CA1.08087.0500 CA1.08087.1000 EM1.08087.0500 EM1.08087.9050 CA1.08087.5000 CA1.08087.9050 EM1.08087.1000
L(+)-Potassium sodium tartrate tetrahydrate, EMSURE® for analysis, Supelco®
L(+)-Potassium sodium tartrate tetrahydrate


Formula: KOOC(CHOH)₂COONa·4H₂O
MW: 282.22 g/mol
Melting Pt: 70…80 °C
Density: ∼1.767 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150989
CAS Number: 6381-59-5
EINECS: 206-156-8
Merck Index: 12,07840
https://us-legacy.vwr.com/stibo/web/std.lang.all/98/18/2299818.jpg
Order Now
Specification Test Results
Solubility 630 g/l (20 °C)
Melting Point 70 - 80 °C
Molar Mass 282.23 g/mol
Bulk Density 1000 kg/m3
Density 7.0 - 8.5 (50 g/l, H2O, 25 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty