Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thioacetamide ACS, Reag. Ph. Eur., GR for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Ethanethioamide

M-Clarity™ quality level = MQ100

1.08170.0250 1.08170.0050 1.08170.0050 1.08170.0250
EM1.08170.0250CS 8784 USD InStock
EM1.08170.0250 EM1.08170.0050 CA1.08170.0250 CA1.08170.0050
Thioacetamide ACS, Reag. Ph. Eur., GR for analysis, Supelco®
Thioacetamide


Formula: CH₃CSNH₂
MW: 75.13 g/mol
Melting Pt: 113…114 °C
Density: 1.37 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008070
CAS Number: 62-55-5
EINECS: 200-541-4
Merck Index: 12,09453
https://us-legacy.vwr.com/stibo/web/std.lang.all/72/66/2297266.jpg
Order Now
Specification Test Results
Assay (argentometric) ≥ 99.0 %
Identity (IR-spectrum) passes test
Melting point 111 - 114 °C
Heavy metals (as Pb) ≤ 0.001 %
Fe (Iron) ≤ 0.0005 %
Sulfated ash ≤ 0.1 %

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty