Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA disodium salt dihydrate, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Ethylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateEthylenediaminetetraacetic acid, disodium salt, dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIKestranal® 2S dihydrateEdetic acid disodium salt dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrate

1.08454.9025 1.08454.0250 1.08454.9025 1.08454.1000 1.08454.0250 1.08454.1000
EM1.08454.0250CS 2915 USD InStock
CA1.08454.0250 CA1.08454.9025 CA1.08454.1000 EM1.08454.0250 EM1.08454.9025 EM1.08454.1000
EDTA disodium salt dihydrate, Supelco®
EDTA disodium salt dihydrate


Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255 °C
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://us-legacy.vwr.com/stibo/web/std.lang.all/99/56/2299956.jpg
Order Now
Specification Test Results
Solubility 100 g/l (20 °C)
Melting Point 110 °C Elimination of water of crystallisation
Molar Mass 372.24 g/mol
Bulk Density 700 kg/m3
pH 4 - 5 (50 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty