Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Xylenol orange tetrasodium salt indicator for metal titration, Supelco®

Supplier: MilliporeSigma
Synonyms: Xylenol orange water soluble

1.08677.0005 1.08677.0005 1.08677.0001 1.08677.0001
EM1.08677.0001CS 19559 USD InStock
EM1.08677.0001 CA1.08677.0005 CA1.08677.0001 EM1.08677.0005
Xylenol orange tetrasodium salt indicator for metal titration, Supelco®
Xylenol orange tetrasodium salt


Formula: C₃₁H₂₈N₂Na₄O₁₃S
MW: 760.59 g/mol
Melting Pt: ∼210 °C
Storage Temperature: Ambient
MDL Number: MFCD00044293
CAS Number: 3618-43-7
EINECS: 222-805-8
Order Now
Specification Test Results
Solubility 510 g/l (20 °C)
Molar Mass 760.59 g/mol
Bulk Density 250 kg/m3
pH 8.1 (10 g/l, H2O, 20 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty