Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc oxide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Zinci oxidumZinc (II) oxide

1.08849.9025 1.08849.1000 1.08849.0500 1.08849.1000 1.08849.9025 1.08849.0500
EM1.08849.1000CS 3400 USD InStock
EM1.08849.1000 CA1.08849.0500 EM1.08849.9025 EM1.08849.0500 CA1.08849.1000 CA1.08849.9025
Zinc oxide, EMSURE® for analysis, Supelco®
Zinc oxide


Formula: ZnO
MW: 81.39 g/mol
MDL Number: MFCD00011300
CAS Number: 1314-13-2
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Solubility (20 °C) insoluble
Melting Point 1975 °C
Molar Mass 81.37 g/mol
Bulk Density 200 - 700 kg/m3
Density 5.61 g/cm3 (20 °C)
pH 7 (50 g/l, H2O, 20 °C) (slurry)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty