Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate heptahydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

M-Clarity™ quality level = MQ300

1.08883.5000 1.08883.5000 1.08883.0500 1.08883.1000 1.08883.1000 1.08883.0500
EM1.08883.0500EA 429 USD InStock
CA1.08883.5000 EM1.08883.0500 CA1.08883.1000 CA1.08883.0500 EM1.08883.1000 EM1.08883.5000
Zinc sulfate heptahydrate, EMSURE® for analysis, Supelco®
Zinc sulphate heptahydrate
EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph.Eur quality specifications.

In North America 1.08883.0500 replaces and has a superior specification to ZX0105-1
In North America 1.08883.9050 replaces and has a superior specification to ZX0105-20


Formula: ZnSO₄·7H₂O
MW: 287.54 g/mol
Melting Pt: 100 °C
Density: 1.97 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149894
CAS Number: 7446-20-0
EINECS: 231-793-3
UN: 3077
ADR: 9,III
Merck Index: 12,10293
Order Now
Specification Test Results
Assay (complexometric) 99.5-103.0%
Ammonium (NH4) 0.001% max
Arsenic (As) 0.00005% max
Cadmium (Cd) 0.0005% max
Calcium (Ca) 0.001% max
Chloride (Cl) 0.0005% max
Clarity of solution passes test
Copper (Cu) 0.0005 max
Identity passes test
Insoluble matter 0.01% max
Iron (Fe) 0.0005% max
Lead (Pb) 0.001 max
Magnesium (Mg) 0.001% max
Manganese (Mn) 0.0003% max
Minimum shelf life passes test
Nitrate (NO3) 0.002% max
Original Examination Date passes
pH (5%, water, 25°C) 4.4-5.6
Potassium (K) 0.001% max
Sodium(Na) 0.001% max
Total nitrogen (N) 0.0005% max

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty