Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Certipur® TOC standard solution acc. to EN 1484-H3/DIN 38409-H3 as potassium hydrogen phthalate in water, stabilized

Supplier: MilliporeSigma
Certipur®
1.09017.0100 1.09017.0100
EM1.09017.0100CS 4731 USD InStock
CA1.09017.0100 EM1.09017.0100
Certipur® TOC standard solution acc. to EN 1484-H3/DIN 38409-H3 as potassium hydrogen phthalate in water, stabilized
Standards Environmental Analysis Standards
  • Diluted standard solutions that are ready-to-use for accurate measurements
  • Traceable to SRM from NIST (SRM 84L)
  • Full certificates of analysis with comprehensive quality documentation
  • Information on concentration and expanded measurement uncertainty
  • Save time and minimize errors

Caution: Store at +15 to +25 °C.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty