Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: Muriatic acidHydrochloric acid solution

M-Clarity™ quality level = MQ200

1.09057.4000 1.09057.4000 1.09057.9025 1.09057.2500 1.09057.9010 1.09057.2500 1.09057.5000 1.09057.9010 1.09057.1000 1.09057.9025 1.09057.5000 1.09057.1000
EM1.09057.2500CS 468 USD InStock
CA1.09057.9025 CA1.09057.5000 CA1.09057.4000 CA1.09057.1000 EM1.09057.2500 EM1.09057.9010 CA1.09057.2500 EM1.09057.5000 CA1.09057.9010 EM1.09057.4000 EM1.09057.1000 EM1.09057.9025
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Hydrochloric acid
In North America 1.09057.1000 is identical to HX0603D-8 but in a larger pack size.


Formula: HCl
MW: 36.46 g/mol
Density: 1.02 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Amount-of-substance concentration 0.995 - 1.005 mol/L
Form liquid
Measurement uncertainty ± 0.003 mol/L

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty