Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulfuric acid 0.25 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Warning

M-Clarity™ quality level = MQ100

1.09073.9010 1.09073.4000 1.09073.9010 1.09073.4000 1.09073.1000 1.09073.1000
EM1.09073.9010EA 322 USD InStock
CA1.09073.4000 EM1.09073.9010 CA1.09073.1000 CA1.09073.9010 EM1.09073.4000 EM1.09073.1000
Sulfuric acid 0.25 mol/l (0.5 N) in aqueous solution, Titripur® volumetric solution, Supelco®
Sulfuric acid


Formula: H₂SO₄
MW: 98.08 g/mol
Density: 1.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 3264
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty