Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Potassium hydrateCaustic potash

M-Clarity™ quality level = MQ100

1.09115.2500 1.09115.1000 1.09115.2500 1.09115.1003
EM1.09115.1000CS 1104 USD InStock
EM1.09115.1000 CA1.09115.2500 EM1.09115.2500 EM1.09115.1003
Potassium hydroxide 0.1 mol/l (0.1 N) in ethanol, Titripur® volumetric solution, Supelco®
Potassium hydroxide
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Formula: KOH
MW: 56.11 g/mol
Density: 0.836 g/cm³ (20 °C)
Flash Pt: 12 °C (EtOH)
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 2924
ADR: 3 (8),II
Order Now
Specification Test Results
Form liquid
Amount-of-substance concentration 0.0995-0.1005
Measurement uncertainty +/- 0.0003
Traceability NIST SRM

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty