Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

M-Clarity™ quality level = MQ100

1.09137.4000 1.09137.1003 1.09137.9010 1.09137.9025 1.09137.4000 1.09137.2500 1.09137.9010 1.09137.1000 1.09137.9025 1.09137.1000 1.09137.2500
EM1.09137.9010EA 228 USD InStock
CA1.09137.1000 EM1.09137.9010 CA1.09137.4000 EM1.09137.1003 EM1.09137.2500 CA1.09137.9025 EM1.09137.1000 CA1.09137.9010 CA1.09137.2500 EM1.09137.4000 EM1.09137.9025
Sodium hydroxide 1 mol/l (1 N) in aqueous solution, Titripur® Reag. Ph. Eur., Reag. USP volumetric solution, Supelco®
Sodium hydroxide
Merck have introduced into the range of Titripur® Titrants some packs with additional technology - Supelco® SmartTitrants that use an RFID tag to provide a seamless data transfer between the smart titrator and the RFID tag on the label of the bottle of titrant. It will ensure that all information needed for method execution, result calculation and documentation is transferred to the titrator equipped with this technology. MettlerToledo Compact and Excellence titrators include this technology. Please see ordering table below for packs with RFID tag.


Formula: NaOH
MW: 40 g/mol
Density: 1.04 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II
Order Now
Specification Test Results
Form liquid
Amount-of-substance concentration 0.995 - 1.005
Measurement uncertainty +/- 0.003
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C.

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty