Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Papanicolaou's solution 3b polychromatic solution EA 50 used in nuclear staining, for cytology, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

1.09272.0500 1.09272.2500 1.09272.1000
EM1.09272.1000EA 122.38 USD InStock
EM1.09272.1000 EM1.09272.0500 EM1.09272.2500
Papanicolaou's solution 3b polychromatic solution EA 50 used in nuclear staining, for cytology, Sigma-Aldrich®
Papanicolaou's solution 3b polychromatic solution EA 50
Used to demonstrate the differentiation of squamous cells in gynecological cytology

The cytological staining technique developed by Papanicolaou is still the standard method in cancer and hormone cycle diagnosis. Four different polychrome solutions are available; they differ in the various concentrations of the components and produce different cytoplasm stains. In the last staining step is used the so-called polychrome solution, a mixture of eosin, light green SF and Bismarck brown. The polychrome solution is used to demonstrate the differentiation of squamous cells. In gynecological cytology Polychrome solution EA 31 (3a) and EA 50 (3b) are mostly used. In non-gynecological cytology Polychrome solution EA 65 (3c) and EA 65 (3d) are used. It is an IVD product and CE certified.


Density: 0.82 g/cm³ (20 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
UN: 1987
ADR: 3,II
IVDLogo
Order Now
Specification Test Results
Suitability for microscopy (Vaginal smear) passes test
Nuclei blue to dark violet
Cyanophilic cytoplasm blue-green
Eosinophilic cytoplasm pink

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty