Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Potassium hydrateCaustic potash

M-Clarity™ quality level = MQ100

1.09351.1000 1.09351.1000
EM1.09351.1000CS 1037 USD InStock
EM1.09351.1000 CA1.09351.1000
Potassium hydroxide 0.5 mol/l (0.5 N) in methanol, Titripur® volumetric solution, Supelco®
Potassium hydroxide


Formula: KOH
MW: 56.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00003553
CAS Number: 1310-58-3
EINECS: 215-181-3
UN: 3286
ADR: 3 (6.1, 8),II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty