Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluenesulfonic acid monohydrate, EMSURE® ACS for analysis, Supelco®

Supplier: MilliporeSigma
Warning
Synonyms: 4-Methylbenzenesulphonic acid hydrate4-Methylbenzenesulfonic acid hydrateToluene-4-sulphonic acid monohydrateToluene-4-sulfonic acid monohydrate

1.09613.0500 1.09613.0100 1.09613.0100 1.09613.0500
EM1.09613.0100CS 3497 USD InStock
EM1.09613.0100 EM1.09613.0500 CA1.09613.0100 CA1.09613.0500
p-Toluenesulfonic acid monohydrate, EMSURE® ACS for analysis, Supelco®
p-Toluenesulphonic acid monohydrate
In North America 1.09613.0500 replaces and is identical to TX0770-3


Formula: C₇H₈O₃S·1H₂O
MW: 190.22 g/mol
Melting Pt: 102…104 °C
Density: 1.24 g/cm³ (20 °C)
Flash Pt: 180 °C
Storage Temperature: Ambient
MDL Number: MFCD00142137
CAS Number: 6192-52-5
EINECS: 203-180-0
UN: 2585
ADR: 8,
Merck Index: 12,09671
Order Now
Specification Test Results
Solubility 670 g/l (20 °C)
Melting Point 56 °C Elimination of water of crystallisation
Molar Mass 190.22 g/mol
Bulk Density 510 kg/m3
Boiling Point 140 °C (27 hPa)
Flash Point 180 °C
pH 1 (650 g/l, H2O, 20 °C) (anhydrous substance)
Ignition Point 600 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty