Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl acetate ≥99.5% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: EtOAcAcetidinEthyl ethanoateVinegar naphthaEthyl acetic esterAcetic acid ethyl esterAcetic etherAcetoxyethane

1.09623.9026 1.09623.2500 1.09623.9026 1.09623.1000 1.09623.9181 1.09623.9181 1.09623.6025 1.09623.1000 1.09623.6025 1.09623.2500
EM1.09623.2500CS 723 USD InStock
CA1.09623.9181 EM1.09623.2500 CA1.09623.1000 CA1.09623.6025 CA1.09623.2500 EM1.09623.6025 CA1.09623.9026 EM1.09623.9181 EM1.09623.9026 EM1.09623.1000
Ethyl acetate ≥99.5% (by GC), EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Ethyl acetate


Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 76.5…77.5 °C (lit.)
Melting Pt: −84 °C (lit.)
Density: 0.902 g/ml (25°C) (lit.)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://us-legacy.vwr.com/stibo/web/std.lang.all/20/72/722072.jpg
Order Now
Specification Test Results
Solubility 85.3 g/l (20 °C)
Melting Point -83 °C
Molar Mass 88.11 g/mol
Boiling Point 77 °C (1013 hPa)
Vapor Pressure 97 hPa (20 °C)
Flash Point -4 °C
Refractive Index 1.37
Explosion Limit 2.1 - 11.5 %(V)
Density 0.90 g/cm3 (20 °C)
Ignition Point 460 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty