Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Cyclohexane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Synonyms: Hexahydro-BenzeneHexamethylene

M-Clarity™ quality level = MQ300

1.09666.2511 1.09666.4000 1.09666.2511 1.09666.4000
EM1.09666.4000CS 2019 USD InStock
EM1.09666.4000 EM1.09666.2511 CA1.09666.4000 CA1.09666.2511
Cyclohexane, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Cyclohexane


Formula: C₆H₁₂
MW: 84.16 g/mol
Melting Pt: 7 °C
MDL Number: MFCD00003814
CAS Number: 110-82-7
UN: 1145
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Solubility 55 mg/l (20°C)
Melting Point 6 °C
Boiling Point 81 °C (1013 hPa)
Vapor Pressure 103 hPa (20 °C)
Flash Point -18 °C
Refractive Index 1.4264 (20 °C)
Explosion Limit 1.2 - 8.3 %(V)
Density 0.78 g/cm3 (20 °C)
pH (H2O) not applicable
Ignition Point 260°C
Viscosity Kinematic 1.26 mm2/s (20 °C)
Grade ACS,ISO,Reag. Ph Eur
Synonyms Hexahydrobenzene, Hexamethylene, Naphthene
Hill Formula C6H12

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty