Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formamide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Carbamaldehyde

1.09684.1000 1.09684.2500 1.09684.1000 1.09684.2500
EM1.09684.1000CS 3038 USD InStock
CA1.09684.2500 EM1.09684.1000 CA1.09684.1000 EM1.09684.2500
Formamide, EMSURE® ACS, Reag. Ph. Eur. for analysis, Supelco®
Formamide


Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://us-legacy.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point 2.9 °C
Molar Mass 45.04 g/mol
Boiling Point 210 °C (1013 hPa)
Vapor Pressure 0.08 hPa (20 °C)
Flash Point 175 °C
Explosion Limit 2.7 - 19.0 %(V)
Density 1.13 g/cm3 (20 °C)
pH 8 - 10 (200 g/l, H2O, 20 °C)
Ignition Point 500 °C

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty