Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

n-Octane reference substance for gas chromatography, Supelco®

Supplier: MilliporeSigma
Danger

1.09716.0005 1.09716.0005
EM1.09716.0005CS 83538 USD InStock
EM1.09716.0005 CA1.09716.0005
n-Octane reference substance for gas chromatography, Supelco®
n-Octane


Formula: H₃C(CH₂)₆CH₃
MW: 114.23 g/mol
Boiling Pt: 126 °C (1013 hPa)
Melting Pt: –57 °C
Density: 0.703 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00009556
CAS Number: 111-65-9
EINECS: 203-892-1
UN: 1262
ADR: 3,
Merck Index: 12,06847
https://us-legacy.vwr.com/stibo/web/std.lang.all/79/71/737971.jpg
Order Now
Specification Test Results
Solubility 0.0007 g/l (20 °C)
Melting Point -57 °C
Molar Mass 114.23 g/mol
Boiling Point 125 - 126 °C (1013 hPa)
Vapor Pressure 14 hPa (20 °C)
Refractive Index 13 °C
Explosion Limit 0.8 - 6.5 %(V)
Density 0.70 g/cm3 (20 °C)
Ignition Point 210 °C
Saturation concentration 70 g/m3 (20 °C) Air
Hazard Class 3
Hazard Class Description Flammable liquid

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty