Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran ≥99.8% (by GC), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger
Synonyms: Butylene oxideOxolaneTHFCyclotetramethylene oxideTetramethylene oxideDiethylene oxide1,4-epoxy-ButaneButane alpha,delta-oxideOxacyclopentane

1.09731.2500 1.09731.6010 1.09731.6025 1.09731.4000 1.09731.6010 1.09731.1000 1.09731.2500 1.09731.1000 1.09731.4000 1.09731.6025
EM1.09731.1000CS 1024 USD InStock
EM1.09731.1000 CA1.09731.6025 EM1.09731.4000 CA1.09731.2500 EM1.09731.6010 CA1.09731.1000 EM1.09731.2500 EM1.09731.6025 CA1.09731.6010 CA1.09731.4000
Tetrahydrofuran ≥99.8% (by GC), EMSURE® for analysis, Supelco®
Tetrahydrofuran


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C (1013 hPa)
Melting Pt: –108.5 °C
Density: 0.889 g/ml (25°C) (lit.)
Flash Pt: –21.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,
REACH: 05-2114615188-48
Merck Index: 12,09351
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now
Specification Test Results
Solubility (20 °C) soluble
Melting Point -108.5 °C
Molar Mass 72.11 g/mol
Boiling Point 65 - 66 °C
Vapor Pressure 173 hPa (20 °C)
Flash Point -21.5 °C
Refractive Index 1.407 (20 °C)
Explosion Limit 1.5 - 12.4 %(V)
Density 0.89 g/cm3 (20 °C)
pH 7 - 8 (200 g/l, H2O, 20 °C)
Ignition Point 215 °C
Evaporation number 2.3

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty