Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(Trimethylsilyl)imidazole for gas chromatography, Supelco®

Supplier: MilliporeSigma
Synonyms: TSIM1-(Trimethylsilyl)-1H-imidazoleTMSIN-(Trimethylsilyl)imidazoleN-(Trimethylsilyl)-1H-imidazole

1.09771.0005 1.09771.0005
EM1.09771.0005CS 48447 USD InStock
CA1.09771.0005 EM1.09771.0005
1-(Trimethylsilyl)imidazole for gas chromatography, Supelco®
1-(Trimethylsilyl)imidazole


Formula: C₆H₁₂N₂Si
MW: 140.26 g/mol
Boiling Pt: 209 °C (1013 hPa)
Melting Pt: –42 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: 61 °C
MDL Number: MFCD00005280
CAS Number: 18156-74-6
EINECS: 242-040-3
Order Now
Specification Test Results
Solubility (20 °C) Risk of violent reaction.
Melting Point -42 °C
Molar Mass 140.25 g/mol
Boiling Point 212 °C (decomposition)
Flash Point 61 °C DIN 51755 Part 1
Density 0.95 g/cm3 (25 °C)
Ignition Point 540 °C DIN 51794
Viscosity kinematic 2.5 mm2/s (25 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty