Would you like to log in again?
Bacillus cereus Selective Supplement, EMD Millipore
Supplier: EMD Millipore
1.09875.0010 1.09875.0010
EM1.09875.0010EA 194.84 USD InStock
EM1.09875.0010 CA1.09875.0010
Bacillus cereus Selective Supplement, EMD Millipore
Media for Microbiology Additives and Supplements for Microbiology
This Bacillus cereus Selective Supplement contains polymyxin B sulfate in lyophilized form.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty