Would you like to log in again?
MQuant™ Nitrite Test Strips, MilliporeSigma
Supplier: MilliporeSigma
MQuant™
1.10057.0001 1.10007.0001 1.10022.0001 1.10007.0002 1.10057.0001 1.10007.0002 1.10007.0001 1.10022.0001
EM1.10007.0002CS 14445 USD InStock
CA1.10022.0001 EM1.10007.0002 EM1.10007.0001 EM1.10057.0001 CA1.10007.0002 CA1.10007.0001 EM1.10022.0001 CA1.10057.0001
MQuant™ Nitrite Test Strips, MilliporeSigma
Environmental Test Kits
MQuant™ (MQ) test strips can be considered as a mobile laboratory on a few square millimetres of plastic strip
  • Easy to handle: just dip, wait and measure visually against colour scale
  • Safe: no auxiliary reagents necessary
  • Immediate results within 2 minutes enables fast decisions
  • Portable: can be used inside or outside the laboratory
MilliporeSigma # 1.10022.0001 replaces and is identical to MilliporeSigma # 10022-1. MilliporeSigma # 1.10007.0001 replaces and is identical to MilliporeSigma # 10007-1
As a screening method they provide the user with a rapid overview of the situation, which helps in making the choice of what, and perhaps if, further analysis is required, possibly leading to substantial savings in time and analytical resources. The strip is a biologically degradable polyester foil and the low reagent content in the reaction zones offers problem-free waste disposal.
1.0022.0001 can be used for measuring the nitrite concentration of boiler water and cooling water.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty