Would you like to log in again?
Chromate Test Method: Colorimetric with Test Strips and Reagent 3 - 10 - 30 - 100 mg/L CrO₄ MQuant™
Supplier: MilliporeSigma
1.10012.0001 1.10012.0001
EM1.10012.0001CS 6147 USD InStock
EM1.10012.0001 CA1.10012.0001
Chromate Test Method: Colorimetric with Test Strips and Reagent 3 - 10 - 30 - 100 mg/L CrO₄ MQuant™
Environmental Test Kits
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty